siRNA Details
| Gene Symbol: TM2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032027.2 | NP_114416.1 | 6 | 840 | ||
| NR_135160.1 | 7 | 970 | ||||
| NR_135161.1 | 5 | 766 | ||||
| NR_135162.1 | 7 | 967 | ||||
| GenBank mRNA | AF353990.1 | - | 6 | 840 | ||
| AI168073.1 | - | 5 | 460 | |||
| AK311444.1 | - | 6 | 486 | |||
| AK311705.1 | - | 7 | 617 | |||
| BC029486.1 | - | 6 | 558 | 5271098 | ||
| BC048995.1 | - | 6 | 556 | |||
| BU664262.1 | - | 7 | 611 | |||

TM2D1
Genomic Map