siRNA Details
| Gene Symbol: IFI27L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032036.2 | NP_114425.1 | 3 | 186 | ||
| GenBank mRNA | AF208232.1 | - | 3 | 152 | ||
| AF238978.1 | - | 1 | 20 | |||
| BC022800.2 | - | 1 | 94 | |||
| BC032626.1 | - | 3 | 145 | 5229498 | ||
| BE737110.1 | - | 187 | ||||

IFI27L2
Genomic Map