siRNA Details
| Gene Symbol: IFI27L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032036.2 | NP_114425.1 | 4 | 429 | ||
| GenBank mRNA | AF208232.1 | - | 4 | 395 | ||
| AF238978.1 | - | 2 | 262 | |||
| BC022800.2 | - | 2 | 337 | |||
| BC032626.1 | - | 4 | 388 | 5229498 | ||
| BE737110.1 | - | 430 | ||||

IFI27L2
Genomic Map