siRNA Details
| Gene Symbol: CCDC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032040.4 | NP_114429.2 | 1 | 1757 | ||
| GenBank mRNA | AL136609.1 | - | 1 | 1683 | ||
| BC025243.1 | - | 1 | 1128 | 4553965 | ||
| JN703457.1 | - | 1 | 1679 | |||
| KT894208.1 | - | 1 | 913 | |||

CCDC8
Genomic Map