siRNA Details
| Gene Symbol: CARD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318011.1 | NP_001304940.1 | 2, 3 | 365 | ||
| NM_032310.4 | NP_115686.3 | 2, 3 | 365 | |||
| NR_134465.1 | 2, 3 | 365 | ||||
| GenBank mRNA | AK057716.1 | - | 2, 3 | 271 | ||
| BC004500.2 | - | 3, 4 | 234 | 3833318 | ||
| BC038856.1 | - | 2, 3 | 258 | 5215640 | ||
| BM930683.1 | - | 2, 3 | 210 | |||

CARD19
Genomic Map