siRNA Details
| Gene Symbol: WDR83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099737.2 | NP_001093207.1 | 3 | 435 | ||
| NM_032332.3 | NP_115708.1 | 1 | 178 | |||
| NR_029375.1 | 3 | 435 | ||||
| GenBank mRNA | AK074525.1 | - | 3 | 425 | ||
| AK223195.1 | - | 1 | 128 | |||
| AK314164.1 | - | 3 | 412 | |||
| BC005870.2 | - | 1 | 111 | 2961616 | ||
| CB143206.1 | - | 1 | 178 | |||

WDR83
Genomic Map