siRNA Details
| Gene Symbol: MEDAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032849.3 | NP_116238.2 | 4 | 921 | ||
| XM_017020801.1 | XP_016876290.1 | 3 | 1301 | |||
| GenBank mRNA | AB055407.1 | - | 3 | 307 | ||
| AK027740.1 | - | 520 | ||||
| AK055635.1 | - | 4 | 905 | |||
| BC040165.1 | - | 4 | 805 | 5590156 | ||
| BC101624.1 | - | 4 | 659 | 8069130 | ||
| BC101626.1 | - | 4 | 659 | 8069132 | ||

MEDAG
Genomic Map