siRNA Details
| Gene Symbol: IFITM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003641.3 | NP_003632.3 | 2 | 572 | ||
| GenBank mRNA | AK290480.1 | - | 2 | 547 | ||
| BC000897.1 | - | 2 | 522 | 3464598 | ||
| J04164.1 | - | 2 | 708 | |||
| X84958.1 | - | 2 | 502 | |||

IFITM1
Genomic Map