siRNA Details
| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003695.2 | NP_003686.1 | 3 | 611 | ||
| GenBank mRNA | BC022806.1 | - | 3 | 538 | ||
| BC031330.1 | - | 3 | 611 | 5428818 | ||
| BC034542.1 | - | 1 | 1232 | |||
| BM019592.1 | - | 3 | 622 | |||
| X82693.1 | - | 3 | 547 | |||

LY6D
Genomic Map