siRNA Details
Gene Symbol: HIST1H2BJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021058.3 | NP_066402.2 | 1 | 203 | ||
GenBank mRNA | AK311849.1 | - | 1 | 231 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HIST1H2BJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021058.3 | NP_066402.2 | 1 | 203 | ||
GenBank mRNA | AK311849.1 | - | 1 | 231 |