siRNA Details
| Gene Symbol: SAMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138352.1 | NP_612361.1 | 1452 | |||
| GenBank mRNA | AY453840.1 | - | 1452 | |||
| BC007384.2 | - | 4, 5 | 546 | |||
| BC030129.2 | - | 4, 5 | 725 | |||
| BC065477.1 | - | 4, 5 | 693 | |||
| BC080588.1 | - | 4, 5 | 546 | |||

SAMD1
Genomic Map