siRNA Details
| Gene Symbol: PKDCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138370.2 | NP_612379.2 | 3 | 1113 | ||
| GenBank mRNA | AK311155.1 | - | 1 | 676 | ||
| BC007901.2 | - | 1 | 68 | |||
| BC062988.1 | - | 1 | 68 | |||
| BC094697.1 | - | 3 | 521 | 30529646 | ||
| BC110513.1 | - | 3 | 550 | 40031755 | ||
| BC110514.1 | - | 3 | 550 | 40031756 | ||
| CF131963.1 | - | 3 | 609 | |||

PKDCC
Genomic Map