siRNA Details
| Gene Symbol: EBAG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278938.1 | NP_001265867.1 | 7 | 651 | ||
| NM_004215.4 | NP_004206.1 | 7 | 938 | |||
| NM_198120.2 | NP_936056.1 | 7 | 771 | |||
| XM_017013960.1 | XP_016869449.1 | 7 | 1134 | |||
| GenBank mRNA | AB007619.1 | - | 7 | 819 | ||
| AF006265.1 | - | 7 | 769 | |||
| AK290651.1 | - | 7 | 760 | |||
| AL515533.3 | - | 7 | 829 | |||
| AY515724.1 | - | 7 | 729 | |||
| AY653072.1 | - | 6, 7 | 671 | |||
| BC005249.1 | - | 7 | 849 | 3930583 | ||
| BC017729.1 | - | 7 | 722 | 3453385 | ||
| BC022506.1 | - | 7 | 836 | 4815654 | ||
| BG616704.1 | - | 7 | 652 | |||
| CR456984.1 | - | 6 | 536 | |||

EBAG9
Genomic Map