siRNA Details
| Gene Symbol: TMSB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021103.3 | NP_066926.1 | 3 | 217 | ||
| GenBank mRNA | AK130949.1 | - | 3 | 205 | ||
| AK311952.1 | - | 3 | 200 | |||
| AY453400.1 | - | 2 | 150 | |||
| BC016025.1 | - | 3 | 182 | 3909431 | ||
| BC016731.1 | - | 3 | 204 | 4095378 | ||
| BC107889.1 | - | 3 | 204 | |||
| M20259.1 | - | 3 | 182 | |||
| M92381.1 | - | 2 | 139 | |||
| S54005.1 | - | 3 | 187 | |||

TMSB10
Genomic Map