siRNA Details
| Gene Symbol: MVB12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304547.1 | NP_001291476.1 | 10 | 1141 | ||
| NM_138401.3 | NP_612410.1 | 9 | 1214 | |||
| GenBank mRNA | AK057057.1 | - | 10 | 1699 | ||
| AK289487.1 | - | 9 | 1014 | |||
| BC007883.2 | - | 10 | 924 | 3688605 | ||
| BC011840.2 | - | 9 | 958 | 4053084 | ||

MVB12A
Genomic Map