siRNA Details
Gene Symbol: MVB12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304547.1 | NP_001291476.1 | 5 | 568 | ||
NM_138401.3 | NP_612410.1 | 4 | 641 | |||
GenBank mRNA | AK057057.1 | - | 5 | 1126 | ||
AK289487.1 | - | 4 | 441 | |||
BC007883.2 | - | 4 | 377 | 3688605 | ||
BC011840.2 | - | 4 | 385 | 4053084 | ||
BG684013.1 | - | 5 | 533 |
