siRNA Details
| Gene Symbol: KBTBD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014867.2 | NP_055682.1 | 2 | 3660 | ||
| XM_011534771.2 | XP_011533073.1 | 2 | 4330 | |||
| XM_011534772.2 | XP_011533074.1 | 3 | 3729 | |||
| XM_017014114.1 | XP_016869603.1 | 4 | 4264 | |||
| XM_017014115.1 | XP_016869604.1 | 3 | 4118 | |||
| XM_017014116.1 | XP_016869605.1 | 3 | 3785 | |||
| XM_017014117.1 | XP_016869606.1 | 2 | 4049 | |||
| GenBank mRNA | AB018254.1 | - | 2 | 3658 | ||

KBTBD11
Genomic Map