siRNA Details
| Gene Symbol: UNC13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080421.2 | NP_001073890.2 | 28 | 3380 | ||
| XM_011527810.2 | XP_011526112.1 | 27 | 3495 | |||
| XM_011527811.2 | XP_011526113.1 | 28 | 3525 | |||
| XM_017026502.1 | XP_016881991.1 | 28 | 3525 | |||
| GenBank mRNA | AB028955.2 | - | 28 | 3378 | ||
| DA236619.1 | - | 3 | 196 | |||

UNC13A
Genomic Map