siRNA Details
| Gene Symbol: PHLPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289003.1 | NP_001275932.1 | 4 | 1198 | ||
| NM_015020.3 | NP_055835.2 | 4 | 851 | |||
| GenBank mRNA | AB023148.2 | - | 4 | 1198 | ||
| AK124678.1 | - | 4 | 522 | |||
| BC129927.1 | - | 3 | 478 | 40124720 | ||

PHLPP2
Genomic Map