siRNA Details
| Gene Symbol: TEX36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128202.2 | NP_001121674.1 | 4 | 673 | ||
| GenBank mRNA | AK131010.1 | - | 3 | 346 | ||
| BC062717.1 | - | 4 | 676 | 6101709 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX36| Gene Symbol: TEX36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128202.2 | NP_001121674.1 | 4 | 673 | ||
| GenBank mRNA | AK131010.1 | - | 3 | 346 | ||
| BC062717.1 | - | 4 | 676 | 6101709 | ||