siRNA Details
| Gene Symbol: C9orf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012715.3 | NP_001012733.2 | 2 | 604 | ||
| GenBank mRNA | AK092588.1 | - | 2 | 605 | ||
| BC132699.1 | - | 2 | 601 | 40146721 | ||
| BC132701.1 | - | 2 | 601 | 40146723 | ||

C9orf106
Genomic Map