siRNA Details
| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109977.1 | NP_001103447.1 | 13 | 3410 | ||
| XM_005263199.1 | XP_005263256.1 | 12 | 3285 | |||
| XM_011532220.1 | XP_011530522.1 | 11 | 3163 | |||
| XM_011532221.2 | XP_011530523.1 | 13 | 3483 | |||
| XM_011532222.2 | XP_011530524.1 | 12 | 3405 | |||
| XM_011532223.2 | XP_011530525.1 | 11 | 3280 | |||
| XM_011532224.2 | XP_011530526.1 | 11 | 2979 | |||
| XM_017008583.1 | XP_016864072.1 | 5 | 1740 | |||
| GenBank mRNA | AK125363.1 | - | 9 | 1643 | ||

FAM160A1
Genomic Map