siRNA Details
| Gene Symbol: TMEM170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100829.2 | NP_001094299.1 | 3 | 333 | ||
| GenBank mRNA | DB092520.1 | - | 3 | 349 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM170B| Gene Symbol: TMEM170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100829.2 | NP_001094299.1 | 3 | 333 | ||
| GenBank mRNA | DB092520.1 | - | 3 | 349 | ||