siRNA Details
Gene Symbol: CLUAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015041.2 | NP_055856.1 | 8 | 964 | ||
NM_024793.2 | NP_079069.1 | 4 | 422 | |||
XM_006720867.3 | XP_006720930.1 | 7 | 815 | |||
XM_017023068.1 | XP_016878557.1 | 8 | 979 | |||
XM_017023069.1 | XP_016878558.1 | 8 | 979 | |||
XM_017023070.1 | XP_016878559.1 | 8 | 964 | |||
XM_017023071.1 | XP_016878560.1 | 8 | 1953 | |||
XM_017023072.1 | XP_016878561.1 | 8 | 1953 | |||
XM_017023073.1 | XP_016878562.1 | 7 | 815 | |||
GenBank mRNA | AB014543.1 | - | 8 | 891 | ||
AB089691.1 | - | 8 | 946 | |||
AK023319.1 | - | 2 | 180 | |||
AK023359.1 | - | 4 | 422 | |||
AK294318.1 | - | 2 | 236 | |||
BC017070.2 | - | 8 | 892 | 3916900 |
