siRNA Details
| Gene Symbol: ZNF609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015042.1 | NP_055857.1 | 2 | 845 | ||
| XM_011521386.2 | XP_011519688.1 | 3 | 1071 | |||
| XM_017022021.1 | XP_016877510.1 | 3 | 1053 | |||
| GenBank mRNA | BC084543.1 | - | 2 | 336 | ||

ZNF609
Genomic Map