siRNA Details
| Gene Symbol: RAB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004161.4 | NP_004152.1 | 5 | 783 | ||
| XM_005264468.1 | XP_005264525.1 | 4 | 492 | |||
| GenBank mRNA | AF498929.1 | - | 5 | 397 | ||
| AK055927.1 | - | 3 | 398 | |||
| AK303678.1 | - | 4 | 491 | |||
| AK315495.1 | - | 5 | 592 | |||
| BC000905.2 | - | 5 | 575 | 2900705 | ||
| BT019528.1 | - | 5 | 397 | |||
| BT019529.1 | - | 5 | 397 | |||
| BX571747.1 | - | 5 | 580 | |||
| CR536488.1 | - | 5 | 397 | |||
| M28209.1 | - | 5 | 447 | |||
| X13898.1 | - | 5 | 397 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008996, AB232583, AF226873, BC002077, AK159425, and others.

RAB1A
Genomic Map