siRNA Details
| Gene Symbol: GSTCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031720.3 | NP_001026890.2 | 8 | 1510 | ||
| NM_024751.3 | NP_079027.2 | 8 | 1431 | |||
| XM_005263222.3 | XP_005263279.1 | 8 | 1698 | |||
| XM_011532248.2 | XP_011530550.1 | 8 | 1654 | |||
| XM_011532249.2 | XP_011530551.1 | 8 | 1516 | |||
| XM_011532252.2 | XP_011530554.1 | 8 | 1888 | |||
| GenBank mRNA | AK292355.1 | - | 8 | 1513 | ||
| AK303547.1 | - | 6 | 777 | |||
| BC032942.2 | - | 8 | 1507 | 4838672 | ||
| BX648355.1 | - | 8 | 1432 | |||

GSTCD
Genomic Map