siRNA Details
| Gene Symbol: CDK11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001313896.1 | NP_001300825.1 | 18 | 2298 | ||
| NM_001313982.1 | NP_001300911.1 | 18 | 2286 | |||
| NM_024011.3 | NP_076916.2 | 18 | 2289 | |||
| NM_033529.3 | NP_277071.2 | 18 | 2259 | |||
| NR_132739.1 | 18 | 2173 | ||||
| NR_132740.1 | 18 | 2212 | ||||
| GenBank mRNA | BC014464.1 | - | 10 | 1020 | ||
| BC094827.1 | - | 10 | 1053 | |||

CDK11A
Genomic Map