siRNA Details
| Gene Symbol: ZNF837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138466.1 | NP_612475.1 | 3 | 1430 | ||
| NR_049780.1 | 3 | 1532 | ||||
| XM_017026242.1 | XP_016881731.1 | 2 | 2198 | |||
| GenBank mRNA | BC012365.1 | - | 2 | 1292 | 4576341 | |
| BC029960.1 | - | 3 | 1533 | |||

ZNF837
Genomic Map