siRNA Details
| Gene Symbol: TBC1D9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015043.3 | NP_055858.2 | 21 | 3653 | ||
| NM_198868.2 | NP_942568.2 | 22 | 3704 | |||
| GenBank mRNA | AB014576.1 | - | 22 | 3704 | ||
| AB449893.1 | - | 22 | 3667 | |||
| AK001196.1 | - | 8 | 1266 | |||
| AK001473.1 | - | 3 | 728 | |||
| AK026096.1 | - | 1 | 215 | |||
| AK074791.1 | - | 11 | 1735 | |||
| AK123874.1 | - | 4 | 2120 | |||
| AK223509.1 | - | 7 | 1204 | |||
| BC000701.2 | - | 8 | 1287 | |||
| BC008919.2 | - | 10 | 1635 | |||
| BC080659.1 | - | 10 | 1714 | |||
| BC110400.1 | - | 12 | 1854 | |||
| BC129994.1 | - | 21 | 3654 | 40129194 | ||
| BX641107.1 | - | 2 | 2370 | |||

TBC1D9B
Genomic Map