siRNA Details
| Gene Symbol: MAST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015183.2 | NP_055998.1 | 3 | 160 | ||
| NM_198828.2 | NP_942123.1 | 5 | 1003 | |||
| XM_011543382.2 | XP_011541684.1 | 6 | 1059 | |||
| XM_017009447.1 | XP_016864936.1 | 6 | 1059 | |||
| GenBank mRNA | AY830839.1 | - | 3 | 161 | ||
| BC033215.1 | - | 5 | 971 | 5015614 | ||

MAST4
Genomic Map