siRNA Details
| Gene Symbol: CCNYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142300.1 | NP_001135772.1 | 5, 6 | 838 | ||
| NM_152523.2 | NP_689736.1 | 8, 9 | 739 | |||
| XM_005246343.1 | XP_005246400.1 | 7, 8 | 757 | |||
| GenBank mRNA | AK095479.1 | - | 2, 3 | 178 | ||
| AK097751.1 | - | 8, 9 | 739 | |||
| BC067253.1 | - | 5, 6 | 625 | 6500304 | ||

CCNYL1
Genomic Map