siRNA Details
| Gene Symbol: CCDC129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257967.1 | NP_001244896.1 | 9 | 1303 | ||
| NM_001257968.1 | NP_001244897.1 | 8 | 1328 | |||
| NM_194300.3 | NP_919276.2 | 7 | 1191 | |||
| XM_011515213.1 | XP_011513515.1 | 8 | 1295 | |||
| XM_017011871.1 | XP_016867360.1 | 8 | 1288 | |||
| XM_017011872.1 | XP_016867361.1 | 8 | 1295 | |||
| GenBank mRNA | AK095663.1 | - | 6 | 1064 | ||
| AK295011.1 | - | 8 | 1246 | |||
| AK304387.1 | - | 9 | 1303 | |||
| BC132719.1 | - | 8 | 1194 | 40146741 | ||

CCDC129
Genomic Map