siRNA Details
| Gene Symbol: CCDC129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257967.1 | NP_001244896.1 | 12 | 2087 | ||
| NM_001257968.1 | NP_001244897.1 | 11 | 2112 | |||
| NM_194300.3 | NP_919276.2 | 10 | 1975 | |||
| NR_047565.1 | 11 | 2489 | ||||
| XM_011515213.1 | XP_011513515.1 | 11 | 2079 | |||
| XM_017011871.1 | XP_016867360.1 | 11 | 2072 | |||
| XM_017011872.1 | XP_016867361.1 | 11 | 2079 | |||
| GenBank mRNA | AK095663.1 | - | 9 | 1848 | ||
| AK128026.1 | - | 11 | 2489 | |||
| AK295011.1 | - | 11 | 2030 | |||
| BC132719.1 | - | 11 | 1978 | 40146741 | ||

CCDC129
Genomic Map