siRNA Details
| Gene Symbol: FXYD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136007.1 | NP_001129479.1 | 11 | 1197 | ||
| NM_001136011.1 | NP_001129483.1 | 10 | 1075 | |||
| NM_001136012.1 | NP_001129484.1 | 8 | 1069 | |||
| NM_005971.3 | NP_005962.1 | 9 | 991 | |||
| NM_021910.2 | NP_068710.1 | 9 | 1153 | |||
| GenBank mRNA | BC090044.1 | - | 9 | 940 | 30395606 | |
| U28249.1 | - | 9 | 1079 | |||
| U31214.1 | - | 1 | 68 | |||

FXYD3
Genomic Map