siRNA Details
| Gene Symbol: C19orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135580.1 | NP_001129052.1 | 1, 2 | 103 | ||
| GenBank mRNA | BX396051.2 | - | 1, 2 | 107 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C19orf71| Gene Symbol: C19orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135580.1 | NP_001129052.1 | 1, 2 | 103 | ||
| GenBank mRNA | BX396051.2 | - | 1, 2 | 107 | ||