siRNA Details
| Gene Symbol: SNX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102575.1 | NP_001096045.1 | 1 | 644 | ||
| NM_001145427.1 | NP_001138899.1 | 1 | 648 | |||
| NM_052870.2 | NP_443102.2 | 1 | 644 | |||
| XM_017008997.1 | XP_016864486.1 | 1 | 648 | |||
| GenBank mRNA | AF395536.1 | - | 1 | 454 | ||
| AK304195.1 | - | 1 | 648 | |||
| BC060791.1 | - | 1 | 644 | 30341956 | ||
| BC067860.1 | - | 1 | 644 | 30343220 | ||
| BC117218.1 | - | 1 | 542 | 40125769 | ||
| BC117220.1 | - | 1 | 542 | 40125771 | ||

SNX18
Genomic Map