siRNA Details
| Gene Symbol: WEE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105558.1 | NP_001099028.1 | 6 | 1309 | ||
| GenBank mRNA | AK131218.1 | - | 6 | 1309 | ||
| BM790836.1 | - | 3 | 228 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WEE2| Gene Symbol: WEE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105558.1 | NP_001099028.1 | 6 | 1309 | ||
| GenBank mRNA | AK131218.1 | - | 6 | 1309 | ||
| BM790836.1 | - | 3 | 228 | |||