siRNA Details
| Gene Symbol: HEATR5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015473.3 | NP_056288.2 | 31 | 5088 | ||
| GenBank mRNA | AB037737.1 | - | 22 | 3536 | ||
| AK000343.1 | - | 9 | 1374 | |||
| AK023789.1 | - | 13 | 2095 | |||
| BC045764.1 | - | 25 | 4101 | |||
| BC063867.1 | - | 13 | 2112 | |||
| CR749444.1 | - | 24 | 3786 | |||

HEATR5A
Genomic Map