siRNA Details
| Gene Symbol: SLC22A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004326.4 | NP_001004326.4 | 1, 2 | 409 | ||
| NR_033396.1 | 1, 2 | 409 | ||||
| XM_011545043.2 | XP_011543345.1 | 1, 2 | 409 | |||
| XM_017017774.1 | XP_016873263.1 | 1, 2 | 409 | |||
| GenBank mRNA | AK131327.1 | - | 1, 2 | 411 | ||
| BC146850.1 | - | 1, 2 | 401 | 9056778 | ||
| DY654625.1 | - | 1, 2 | 51 | |||

SLC22A20
Genomic Map