siRNA Details
Gene Symbol: SLC22A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033396.1 | 9, 10 | 1609 | |||
XM_017017775.1 | XP_016873264.1 | 6, 7 | 1416 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC22A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033396.1 | 9, 10 | 1609 | |||
XM_017017775.1 | XP_016873264.1 | 6, 7 | 1416 |