siRNA Details
| Gene Symbol: YIPF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182592.2 | NP_872398.2 | 3 | 243 | ||
| XM_011513679.2 | XP_011511981.1 | 4 | 5584 | |||
| GenBank mRNA | AK096895.1 | - | 3 | 243 | ||
| BC103996.1 | - | 3 | 243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

YIPF7| Gene Symbol: YIPF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182592.2 | NP_872398.2 | 3 | 243 | ||
| XM_011513679.2 | XP_011511981.1 | 4 | 5584 | |||
| GenBank mRNA | AK096895.1 | - | 3 | 243 | ||
| BC103996.1 | - | 3 | 243 | |||