siRNA Details
| Gene Symbol: YIPF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182592.2 | NP_872398.2 | 4 | 411 | ||
| XM_011513679.2 | XP_011511981.1 | 5 | 5752 | |||
| GenBank mRNA | AK096895.1 | - | 4 | 411 | ||
| BC103995.1 | - | 3 | 247 | |||
| BC103996.1 | - | 4 | 416 | |||

YIPF7
Genomic Map