siRNA Details
| Gene Symbol: POMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013382.5 | NP_037514.2 | 2 | 500 | ||
| XM_011536675.2 | XP_011534977.1 | 2 | 500 | |||
| XM_011536677.2 | XP_011534979.1 | 2 | 501 | |||
| XM_017021244.1 | XP_016876733.1 | 2 | 494 | |||
| GenBank mRNA | AB209114.1 | - | 2 | 322 | ||
| AF105020.1 | - | 2 | 503 | |||
| AL554279.3 | - | 2 | 364 | |||
| AY090480.1 | - | 2 | 503 | |||
| BC031651.1 | - | 2 | 390 | 5169145 | ||
| CR627447.1 | - | 2 | 70 | |||

POMT2
Genomic Map