siRNA Details
| Gene Symbol: DNAJC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307980.1 | NP_001294909.1 | 5 | 972 | ||
| NM_001307981.1 | NP_001294910.1 | 5 | 969 | |||
| NM_005528.3 | NP_005519.2 | 6 | 1008 | |||
| GenBank mRNA | AF036874.1 | - | 5 | 615 | ||
| BC010145.2 | - | 3 | 487 | |||
| BC032137.2 | - | 5 | 646 | |||
| BC044584.1 | - | 6 | 1011 | 4811478 | ||
| BC051008.1 | - | 5 | 491 | |||
| BC053947.1 | - | 6 | 1011 | |||
| BC057849.1 | - | 5 | 841 | 30349314 | ||

DNAJC4
Genomic Map