siRNA Details
| Gene Symbol: CNTLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114395.2 | NP_001107867.1 | 7 | 1204 | ||
| NM_017738.3 | NP_060208.2 | 7 | 1204 | |||
| XM_005251492.1 | XP_005251549.1 | 7 | 1204 | |||
| XM_006716793.3 | XP_006716856.1 | 7 | 1002 | |||
| XM_011517941.2 | XP_011516243.1 | 7 | 1204 | |||
| XM_017014839.1 | XP_016870328.1 | 7 | 1204 | |||
| XM_017014840.1 | XP_016870329.1 | 7 | 1204 | |||
| XM_017014841.1 | XP_016870330.1 | 7 | 1204 | |||
| XM_017014842.1 | XP_016870331.1 | 7 | 1204 | |||
| XM_017014843.1 | XP_016870332.1 | 7 | 1204 | |||
| GenBank mRNA | AK000283.1 | - | 3 | 299 | ||
| AK098502.1 | - | 7 | 1155 | |||
| AK308837.1 | - | 7 | 1159 | |||
| BX647069.1 | - | 7 | 1162 | |||

CNTLN
Genomic Map