siRNA Details
| Gene Symbol: CNTLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114395.2 | NP_001107867.1 | 4 | 661 | ||
| NM_017738.3 | NP_060208.2 | 4 | 661 | |||
| XM_005251492.1 | XP_005251549.1 | 4 | 661 | |||
| XM_006716793.3 | XP_006716856.1 | 4 | 459 | |||
| XM_011517941.2 | XP_011516243.1 | 4 | 661 | |||
| XM_017014839.1 | XP_016870328.1 | 4 | 661 | |||
| XM_017014840.1 | XP_016870329.1 | 4 | 661 | |||
| XM_017014841.1 | XP_016870330.1 | 4 | 661 | |||
| XM_017014842.1 | XP_016870331.1 | 4 | 661 | |||
| XM_017014843.1 | XP_016870332.1 | 4 | 661 | |||
| GenBank mRNA | AK098502.1 | - | 4 | 612 | ||
| AK308837.1 | - | 4 | 616 | |||
| BX647069.1 | - | 4 | 619 | |||

CNTLN
Genomic Map