siRNA Details
| Gene Symbol: C22orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302819.1 | NP_001289748.1 | 1 | 475 | ||
| NM_015372.2 | NP_056187.1 | 2 | 266 | |||
| GenBank mRNA | AJ704765.1 | - | 2 | 217 | ||
| AK294232.1 | - | 1 | 445 | |||
| AK315264.1 | - | 2 | 264 | |||
| AL050256.1 | - | 2 | 254 | |||

C22orf24
Genomic Map