siRNA Details
| Gene Symbol: LCE1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178353.1 | NP_848130.1 | 2 | 871 | ||
| GenBank mRNA | BC038391.1 | - | 2 | 871 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCE1E| Gene Symbol: LCE1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178353.1 | NP_848130.1 | 2 | 871 | ||
| GenBank mRNA | BC038391.1 | - | 2 | 871 | ||