siRNA Details
| Gene Symbol: MMACHC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015506.2 | NP_056321.2 | 4 | 707 | ||
| XM_005270724.4 | XP_005270781.1 | 3 | 466 | |||
| XM_011541204.2 | XP_011539506.1 | 4 | 616 | |||
| GenBank mRNA | AK098537.1 | - | 4 | 707 | ||
| AL080062.1 | - | 4 | 578 | |||
| BC006122.1 | - | 4 | 550 | 3535453 | ||
| BU839693.1 | - | 4 | 562 | |||

MMACHC
Genomic Map